missing translation for 'onlineSavingsMsg'
Learn More

Tantalum powder, -22 mesh, Puratronic™, 99.95% (metals basis), Nb 100ppm, Thermo Scientific Chemicals

56.30 € - 450.00 €

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11315809
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010753.06
5 g
56.30 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11336847
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010753.14
25 g
167.00 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11326847
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010753.22
100 g
450.00 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-25-7
180.95
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]

Specifications

Black to Gray
Gray to black powder
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
180.95
CHEBI:33348
Odorless
Tantalum
7440-25-7
99.98% (Metals basis)
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
23956
-22 Mesh
Powder
Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

missing translation for 'dotInformation' : DOT Class: 4.1; Packaging Group: II

missing translation for 'einecsNumber' : 231-135-5

missing translation for 'rtecsNumber' : WW5505000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only