missing translation for 'onlineSavingsMsg'
Learn More

Tantalum rod, 12.7mm (0.5in) dia, hard, 99.95% (metals basis), Thermo Scientific Chemicals

111.00 € - 1492.00 €

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11385788
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010347.BO
1 cm
111.00 €
1cm
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11395788
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010347.BQ
5 cm
475.00 €
5cm
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11305798
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010347.BU
25 cm
1492.00 €
25cm
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-25-7
180.95
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]

Specifications

7440-25-7
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
23956
99.95%
≈20.4g/cm
(metals basis)
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
180.95
CHEBI:33348
Odorless
Rod
Tantalum rod, 12.7mm (0.5 in.) dia., Hard, Temper: as drawn
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-135-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only