missing translation for 'onlineSavingsMsg'
Learn More
Tantalum tris(diethylamido)-tert-butylimide, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11365707

Tantalum tris(diethylamido)-tert-butylimide, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11365707
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11365707

Brand: Thermo Scientific Alfa Aesar 043419.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Precursor for TaN

Tantalum tris(diethylamido)-tert-butylimide is used as a precursor for tantalum nitride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169896-41-7
Molecular Formula C15H39N4Ta
Molecular Weight (g/mol) 456.45
MDL Number MFCD02684506
InChI Key NXXDVZOFTYVIKX-UHFFFAOYSA-N
Synonym tert-butylimino tris diethylamino tantalum, tantalum tris diethylamido-tert-butylimide, tbtdet, tris diethylamino tert-butylimino tantalum, t-butylimido tris diethylamino tantalum v, tris diethylamido tert-butylimido tantalum v, tris diethylamino tert-butylimino tantalum v, tert-butylimino bis diethylamino tantalio diethylamine, tantalum,tris n-ethylethanaminato 2-methyl-2-propanaminato 2--, t-4
PubChem CID 4100858
IUPAC Name tert-butyliminotantalum;diethylazanide
SMILES CCN(CC)[Ta](N(CC)CC)(N(CC)CC)=[N](C)(C)C

Specifications

Color Yellow
Boiling Point 90°C (0.1 mmHg)
Packaging Ampule
Quantity 5 g
UN Number UN3129
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 468.48
Percent Purity 99.99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tantalum tris (diethylamido)-tert-butylimide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tantalum tris(diethylamido)-tert-butylimide, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.