Learn More
Tantalum(V) bromide, 99.9% (metals basis), Thermo Scientific Chemicals
41.81 € - 1630.11 €
Chemical Identifiers
CAS | 13451-11-1 |
---|---|
Molecular Formula | Br5Ta |
Molecular Weight (g/mol) | 580.468 |
MDL Number | MFCD00049563 |
InChI Key | GCPVYIPZZUPXPB-UHFFFAOYSA-I |
Synonym | tantalum pentabromide, tantalum bromide tabr5, tantalum v bromide, tantalum 5+ pentabromide, tabr5 |
PubChem CID | 83480 |
IUPAC Name | pentabromotantalum |
SMILES | Br[Ta](Br)(Br)(Br)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11341579
|
Thermo Scientific Alfa Aesar
014041.04 |
2 g |
41.81 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357657
|
Thermo Scientific Alfa Aesar
014041.09 |
10 g |
126.63 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367657
|
Thermo Scientific Alfa Aesar
014041.18 |
50 g |
454.26 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11331579
|
Thermo Scientific Alfa Aesar
014041.30 |
250 g |
1630.11 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Tantalum Bromide Cluster is used for the preparation of heavy-atom derivatives for structure determination of biological macromolecules by X-ray analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13451-11-1 | |
580.468 | |
GCPVYIPZZUPXPB-UHFFFAOYSA-I | |
83480 | |
Br[Ta](Br)(Br)(Br)Br |
Br5Ta | |
MFCD00049563 | |
tantalum pentabromide, tantalum bromide tabr5, tantalum v bromide, tantalum 5+ pentabromide, tabr5 | |
pentabromotantalum |
Specifications
348.8°C | |
13451-11-1 | |
Powder | |
MFCD00049563 | |
tantalum pentabromide, tantalum bromide tabr5, tantalum v bromide, tantalum 5+ pentabromide, tabr5 | |
Br[Ta](Br)(Br)(Br)Br | |
580.468 | |
580.49 | |
Pungent | |
4.67g/mL | |
Tantalum(V) bromide |
265°C | |
Yellow | |
Br5Ta | |
UN3260 | |
GCPVYIPZZUPXPB-UHFFFAOYSA-I | |
pentabromotantalum | |
83480 | |
99.9% | |
Moisture Sensitive | |
(metals basis) |
Sicherheit und Handhabung
GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.