missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) chloride, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11357757
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

In chlorination of organic substances, as an intermediate, and in the production of pure metalTantalum(V) chloride is used in the preparation of catalyst for the polycyclotrimerizations of alkenediynes, chloro-aryloxide compounds and for the plasma-enhanced atomic layer deposition of tantalum nitride films. It acts as a starting material for new edge-bridged octahedral M6 cluster compounds. It is involved in the preparation of tantalum(V) oxychloride, tantalum pentoxide. It is electrophilic and behaves like a Friedel-Crafts type catalyst, similar to aluminum(III) chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In chlorination of organic substances, as an intermediate, and in the production of pure metal
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7721-01-9
Molecular Formula Cl5Ta
Molecular Weight (g/mol) 358.20
MDL Number MFCD00011253
InChI Key OEIMLTQPLAGXMX-UHFFFAOYSA-I
Synonym Tantalum pentachloride
IUPAC Name tantalum(5+) pentachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]

Specifications

Boiling Point 242°C
Melting Point 216°C
Color White to Yellow
Physical Form Powder
Quantity 5 g
UN Number UN3260
Merck Index 14,9054
Solubility Information Decomposes in water; Soluble in absolute alcohol and KOH; Soluble in ether,carbon tetrachloride,sulfuric acid and potassium hydroxide; Slightly soluble in ethanol.
Formula Weight 358.21
Percent Purity 99.8%
Odor Acidic
Packaging Ampule
Sensitivity Moisture sensitive
Density 3.68 g/mL
Assay (metals basis)
Chemical Name or Material Tantalum(V) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'