Learn More
Tantalum(V) chloride, 99.8% (metals basis), Thermo Scientific Chemicals
Description
In chlorination of organic substances, as an intermediate, and in the production of pure metalTantalum(V) chloride is used in the preparation of catalyst for the polycyclotrimerizations of alkenediynes, chloro-aryloxide compounds and for the plasma-enhanced atomic layer deposition of tantalum nitride films. It acts as a starting material for new edge-bridged octahedral M6 cluster compounds. It is involved in the preparation of tantalum(V) oxychloride, tantalum pentoxide. It is electrophilic and behaves like a Friedel-Crafts type catalyst, similar to aluminum(III) chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- In chlorination of organic substances, as an intermediate, and in the production of pure metal

Specifications
Specifications
Boiling Point | 242°C |
Melting Point | 216°C |
Color | White to Yellow |
Physical Form | Powder |
Quantity | 25 g |
UN Number | UN3260 |
Merck Index | 14,9054 |
Solubility Information | Decomposes in water; Soluble in absolute alcohol and KOH; Soluble in ether,carbon tetrachloride,sulfuric acid and potassium hydroxide; Slightly soluble in ethanol. |
Formula Weight | 358.21 |
Percent Purity | 99.8% |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.