Learn More
Tantalum(V) chloride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals
67.30 € - 483.00 €
Chemical Identifiers
CAS | 7721-01-9 |
---|---|
Molecular Formula | Cl5Ta |
Molecular Weight (g/mol) | 358.20 |
MDL Number | MFCD00011253 |
InChI Key | OEIMLTQPLAGXMX-UHFFFAOYSA-I |
IUPAC Name | tantalum(5+) pentachloride |
SMILES | [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11377018
|
Thermo Scientific Alfa Aesar
014708.06 |
5 g |
67.30 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387018
|
Thermo Scientific Alfa Aesar
014708.14 |
25 g |
177.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367018
|
Thermo Scientific Alfa Aesar
014708.22 |
100 g |
483.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tantalum(V) chloride is used in the preparation of catalyst for the polycyclotrimerizations of alkenediynes, chloro-aryloxide compounds and for the plasma-enhanced atomic layer deposition of tantalum nitride films. It acts as a starting material for new edge-bridged octahedral M6 cluster compounds. It is involved in the preparation of tantalum(V) oxychloride, tantalum pentoxide. It is electrophilic and behaves like a Friedel-Crafts type catalyst, similar to aluminum(III) chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
7721-01-9 | |
358.20 | |
OEIMLTQPLAGXMX-UHFFFAOYSA-I | |
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5] |
Cl5Ta | |
MFCD00011253 | |
tantalum(5+) pentachloride |
Specifications
242°C | |
7721-01-9 | |
Powder | |
MFCD00011253 | |
14,9054 | |
OEIMLTQPLAGXMX-UHFFFAOYSA-I | |
tantalum(5+) pentachloride | |
358.21 | |
Puratronic™ | |
Ampule | |
3.68 g/mL | |
Tantalum(V) chloride |
216°C | |
White to Yellow | |
Cl5Ta | |
UN3260 | |
Soluble in ether,carbon tetrachloride,sulfuric acid and potassium hydroxide. Slightly soluble in ethanol. | |
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5] | |
358.20 | |
99.99% | |
Acidic | |
Moisture sensitive | |
(metals basis) |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 231-755-6
missing translation for 'rtecsNumber' : WW5600000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only