missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) chloride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

67.84 € - 473.36 €

Chemical Identifiers

CAS 7721-01-9
Molecular Formula Cl5Ta
Molecular Weight (g/mol) 358.20
MDL Number MFCD00011253
InChI Key OEIMLTQPLAGXMX-UHFFFAOYSA-I
IUPAC Name tantalum(5+) pentachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11397047
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010880.06
5 g
67.84 €
5g
Please to purchase this item. Need a web account? Register with us today!
11317057
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010880.14
25 g
176.88 €
25g
Please to purchase this item. Need a web account? Register with us today!
11307057
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010880.22
100 g
473.36 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tantalum(V) chloride is used to form mixed chloro-aryloxide compounds, such as [Ta(OC6H3-2,6-i-Pr2)2Cl3]2, which form pyridine or phosphine adducts with a distorted six-coordinate geometry. It is also used as a starting material for new edge-bridged octahedral M6 cluster compounds such as CsErTa6Cl18.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7721-01-9
358.20
OEIMLTQPLAGXMX-UHFFFAOYSA-I
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]
Cl5Ta
MFCD00011253
tantalum(5+) pentachloride

Specifications

242°C
7721-01-9
99.99%, metal basis
MFCD00011253
OEIMLTQPLAGXMX-UHFFFAOYSA-I
tantalum(5+) pentachloride
358.21
3.68
216°C
Resublimed Crystalline
Cl5Ta
Soluble in water (reacts). Also soluble in ethanol,ether and carbon tetrachloride.
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]
358.20
Moisture sensitive
Tantalum(V) Chloride, Ampouled under argon
Videos
Safety and Handling

Safety and Handling

When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

missing translation for 'dotInformation' : DOT Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 231-755-6

missing translation for 'rtecsNumber' : WW5600000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only