missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) ethoxide, 99.999% (metals basis), Nb <100ppm, Thermo Scientific Chemicals

63.72 € - 768.83 €

Chemical Identifiers

CAS 6074-84-6
Molecular Formula C10H25O5Ta
Molecular Weight (g/mol) 406.25
MDL Number MFCD00049785
InChI Key HSXKFDGTKKAEHL-UHFFFAOYSA-N
Synonym tantalum v ethoxide, tantalum 5+ ethanolate, tantalum ethoxide, tantalum 5+ pentaethanolate, ethanol, tantalum 5+ salt, pentaethoxytantalum v, ethanol, tantalum 5+ salt 5:1, tantalum ethylate, pentaethyl tantalate, tantalum pentaethoxide
PubChem CID 160806
IUPAC Name ethanolate;tantalum(5+)
SMILES CCO[Ta](OCC)(OCC)(OCC)OCC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11376958
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014643.03
1 g
63.72 €
1g
Please to purchase this item. Need a web account? Register with us today!
11396958
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014643.09
10 g
194.97 €
10g
Please to purchase this item. Need a web account? Register with us today!
11386958
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014643.18
50 g
768.83 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tantalum(V) ethoxide acts as a precursor used in the preparation of ultra thin films of tantalum oxide and other tantalum containing films, which finds application in semiconductors. It is also used to prepare tantalum oxide nanoparticles for the imaging cartilage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6074-84-6
406.25
HSXKFDGTKKAEHL-UHFFFAOYSA-N
160806
CCO[Ta](OCC)(OCC)(OCC)OCC
C10H25O5Ta
MFCD00049785
tantalum v ethoxide, tantalum 5+ ethanolate, tantalum ethoxide, tantalum 5+ pentaethanolate, ethanol, tantalum 5+ salt, pentaethoxytantalum v, ethanol, tantalum 5+ salt 5:1, tantalum ethylate, pentaethyl tantalate, tantalum pentaethoxide
ethanolate;tantalum(5+)

Specifications

6074-84-6
1.566 g/mL
30°C (87°F)
Ta(OCH2CH3)5
MFCD00049785
3678999
tantalum v ethoxide, tantalum 5+ ethanolate, tantalum ethoxide, tantalum 5+ pentaethanolate, ethanol, tantalum 5+ salt, pentaethoxytantalum v, ethanol, tantalum 5+ salt 5:1, tantalum ethylate, pentaethyl tantalate, tantalum pentaethoxide
HSXKFDGTKKAEHL-UHFFFAOYSA-N
ethanolate;tantalum(5+)
160806
99.999%
Liquid
Tantalum(V) ethoxide
18°C to 19°C
145°C (0.1 mmHg)
C10H25O5Ta
1.488
UN2924
Air and moisture sensitive
Decomposes in water; Soluble in organic solvents.
CCO[Ta](OCC)(OCC)(OCC)OCC
406.25
406.25
Nb <100ppm
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 228-010-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only