Learn More
Tantalum(V) fluoride, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Tantalum(V) fluoride is used as a Friedel-Crafts catalyst. It combines with hydrogen fluoride and acts as a catalyst for the alkylation of alkanes and alkenes as well as the protonation of aromatic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 229.5°C |
Melting Point | 96.8°C |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 99.9%, metal basis |
Solubility Information | Decomposes in water forming tantalum oxyfluoride. Soluble in conc. and fuming HNO3Soluble in carbon disulfide,carbon tetrachloride and chloroform. Slightly soluble in acetic acid and ethanol. |
Formula Weight | 275.94 |
Sensitivity | Moisture sensitive |
Decomposition Information | Decomposes in water forming tantalum oxyfluoride |
Density | 4.74 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y14J007 | Certificate of Analysis | 05/12/2023 | 014052.09,014052.18 |
X27H006 | Certificate of Analysis | 12/04/2022 | 014052.09,014052.18 |
E31L11 | Certificate of Analysis | 07/12/2021 | 014052.09,014052.18 |
M26C010 | Certificate of Analysis | 07/12/2021 | 014052.09,014052.18 |
I05L05 | Certificate of Analysis | 07/12/2021 | 014052.09,014052.18 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.