missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) methoxide, Thermo Scientific Chemicals

Product Code. 11323497
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tantalum(V) methoxide is used in the synthesis of tantalum hydride. Silica-supported alkoxo-tantalum complexes were used in the deperoxidation of cyclohexyl hydroperoxide into cyclohexanol and cyclohexanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-35-0
Molecular Formula C5H15O5Ta
Molecular Weight (g/mol) 336.118
MDL Number MFCD00058817
InChI Key QASMZJKUEABJNR-UHFFFAOYSA-N
PubChem CID 12787641
IUPAC Name methanolate;tantalum(5+)
SMILES C[O-].C[O-].C[O-].C[O-].C[O-].[Ta+5]

Specifications

Melting Point 50°C
Boiling Point 189°C (10 mmHg)
Linear Formula Ta(OCH3)5
Quantity 5 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 336.12
Physical Form Crystalline
Chemical Name or Material Tantalum(V) methoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.