missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) oxide, 99.85% (metals basis), Thermo Scientific Chemicals

Product Code. 11381099
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Tantalum(V) oxide is used in the production of capacitors, due to its high dielectric constant. These capacitors are used in automotive electronics, cell phones, thin film components and electronic circuitry. In addition, Tantalum(V) oxide has been utilized in the fabrication of the glass of photographic lenses. Due to its high biocompatibility and resistivity, it has attracted many biomedical applications. It is used in tantalum capacitors, resistive switching memories, high frequency CMOS (Complementary metal-oxide-semiconductors) integrated circuits and high-k dielectric for DRAM (Dynamic random-access memory) capacitor applications. It has been shown to improve corrosion resistance and cytocompatibility of titanium substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-61-0
Molecular Formula O5Ta2
Molecular Weight (g/mol) 441.89
MDL Number MFCD00011254
InChI Key PBCFLUZVCVVTBY-UHFFFAOYSA-N
IUPAC Name [(dioxotantalio)oxy]dioxotantalum
SMILES O=[Ta](=O)O[Ta](=O)=O

Specifications

Melting Point 1800°C
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.85% (metals basis)
Solubility Information Insoluble in almost all organic solvents. Solubility is negligible in water.
Formula Weight 441.89
Density 8.2
Chemical Name or Material Tantalum(V) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'