missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) oxide, 99.95% (metals basis excluding Nb), Nb <200ppm, Thermo Scientific Chemicals

Product Code. 11343428
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tantalum (V) oxide (Ta2O5) has potential applications as part of an environmental barrier coating system for Si3N4-based turbine components. Octadecyl phosphoric acid ester has been found to produce oriented, well-ordered monolayers on a flat tantalum (V) oxide surface, via self-assembly from a heptane/propan-2-ol solution. Tantalum(V) oxide (Ta 2 O 5 ) has been used extensively as an insulating layer in discrete capacitors and shows a high potential for semiconductor applications such as an insulating dielectric in the capacitive element in memory devices such as dynamic random access.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-61-0
Molecular Formula O5Ta2
Molecular Weight (g/mol) 441.89
MDL Number MFCD00011254
InChI Key PBCFLUZVCVVTBY-UHFFFAOYSA-N
IUPAC Name [(dioxotantalio)oxy]dioxotantalum
SMILES O=[Ta](=O)O[Ta](=O)=O

Specifications

Melting Point 1800°C
Physical Form Sintered Lump
Quantity 100 g
Merck Index 14,9056
Solubility Information Insoluble in water.
Formula Weight 441.89
Percent Purity 99.95%
Odor Odorless
Density 8.2 g/mL
Assay (metals basis excluding Nb), Nb <200ppm
Chemical Name or Material Tantalum(V) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'