missing translation for 'onlineSavingsMsg'
Learn More

Tantalum wire, 0.25mm (0.01in) dia, annealed, 99.9+% (metals basis), Thermo Scientific Chemicals

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
IUPAC Name tantalum
SMILES [Ta]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11325798
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010348.G6
10 m N/A
N/A
11315798
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010348.H2
50 m N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-25-7
180.95
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]

Specifications

7440-25-7
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
≥99.9%
Tantalum wire
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
180.95
Tube
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-135-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only