missing translation for 'onlineSavingsMsg'
Learn More

Ley Rubber™ Tapered Cork Stoppers

Tapered cork stoppers for use in education, homebrewing and arts and crafts.

Brand:  Ley Rubber™ CB018/S/Z

 View more versions of this product

Product Code. 12396457


Description

Description

  • Range of sizes available
  • Recyclable product
Specifications

Specifications

Education, homebrewing, arts and crafts
10/Pk.
21 mm
Cork Stopper
32 mm
18 mm
Cork
Tapered
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ley Rubber™ Tapered Cork Stoppers > Diameter (Metric) Bottom: 18mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.