missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TAPS, 99+%, for biochemistry

Product Code. 10716623
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29915-38-6
Molecular Formula C7H17NO6S
Molecular Weight (g/mol) 243.27
MDL Number MFCD00007538
InChI Key YNLCVAQJIKOXER-UHFFFAOYSA-N
Synonym 3-[Tris(hydroxymethyl)methylamino]-1-propanesulfonic acid
PubChem CID 121591
IUPAC Name 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid
SMILES OCC(CO)(CO)NCCCS(O)(=O)=O

Specifications

Absorbance (0.1 M aq. soln.)
Absorbance Conditions (0.1M aq. soln.)
Melting Point 240.0°C
Color White
pH 5 to 6.5 (1% aq. soln. at 25°C)
Physical Form Powder
Quantity 25 g
Flash Point >110°C
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula (HOCH2)3CNH(CH2)3SO3H
Packaging Glass bottle
Merck Index 15, 9191
Solubility Information Solubility in water: ca 121.6g/L (20°C).
Formula Weight 243.27
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material TAPS
Show More Show Less
Product Identifier
  • TAPS

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'