missing translation for 'onlineSavingsMsg'
Learn More

TAPS (Free Acid) (White Powder), Fisher BioReagents

Product Code. 10502965
Click to view available options
Poly Bottle; 25g
This item is not returnable. View return policy
This item is not returnable. View return policy
This biological buffer has a usable pH range of 7.7 to 9.1.
TRUSTED_SUSTAINABILITY

Specifications

CAS 29915-38-6
Color White
Physical Form Powder
Quantity 25 g
DNase Not Detected
Assay Percent Range ≥99 %
Molecular Formula C7H17NO6S
MDL Number MFCD00007538
Packaging Poly Bottle
Protease Not Detected
Merck Index 15, 9191
Synonym Tris(hydroxymethyl) methylaminopropanesulfonic Acid, 3-[[2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl]amino]-1-Propane-sulfonic Acid
InChI Key YNLCVAQJIKOXER-UHFFFAOYSA-N
SMILES OCC(CO)(CO)NCCCS(O)(=O)=O
IUPAC Name 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid
Molecular Weight (g/mol) 243.27
PubChem CID 121591
Formula Weight 243.18
Percent Purity ≥99%
Chemical Name or Material TAPS (Free Acid)
Show More Show Less
Product Identifier
  • TAPS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.