missing translation for 'onlineSavingsMsg'
Learn More

TAPS, JustPURE™, white powder, Fisher BioReagents

Product Code. 10334830
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Absorbance 0.05 max. at 260nm
CAS 29915-38-6
Melting Point 240°C
Color White
Physical Form Solid
Quantity 100 g
Assay Percent Range ≥99.5 %
Molecular Formula C7H17NO6S
MDL Number MFCD00007538
Packaging Poly Bottle
Merck Index 15, 9191
Synonym [(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)amino]-1-propanesulfonic acid, N-[Tris(hydroxymethyl)methyl]-3-aminopropanesulfonic acid
InChI Key YNLCVAQJIKOXER-UHFFFAOYSA-N
SMILES OCC(CO)(CO)NCCCS(O)(=O)=O
IUPAC Name 3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propane-1-sulfonic acid
Molecular Weight (g/mol) 243.27
PubChem CID 121591
Formula Weight 243.28
Percent Purity ≥99.5%
Grade JustPURE™
Chemical Name or Material TAPS
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'