Learn More
Thermo Scientific Chemicals TAPSO, 99+%, for biochemistry
30.70 € - 240.00 €
Chemical Identifiers
CAS | 68399-81-5 |
---|---|
Molecular Formula | C7H17NO7S |
Molecular Weight (g/mol) | 259.273 |
MDL Number | MFCD00038352 |
InChI Key | RZQXOGQSPBYUKH-UHFFFAOYSA-N |
Synonym | 3-(N-Tris-(hydroxymethyl)methylamino)-2-hydroxypropanesulfonic acid |
PubChem CID | 109334 |
IUPAC Name | 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]-2-hydroxypropane-1-sulfonic acid |
SMILES | C(C(CS(=O)(=O)O)O)NC(CO)(CO)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
13564990
|
Thermo Scientific Chemicals
327810250 |
25 g | Glass bottle |
30.70 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13534990
|
Thermo Scientific Chemicals
327811000 |
100 g | Glass bottle |
81.10 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13594990
|
Thermo Scientific Chemicals
327815000 |
500 g | Glass bottle |
240.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
68399-81-5 | |
259.273 | |
RZQXOGQSPBYUKH-UHFFFAOYSA-N | |
109334 | |
C(C(CS(=O)(=O)O)O)NC(CO)(CO)CO |
C7H17NO7S | |
MFCD00038352 | |
3-(N-Tris-(hydroxymethyl)methylamino)-2-hydroxypropanesulfonic acid | |
3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]-2-hydroxypropane-1-sulfonic acid |
Specifications
(0.1 M aq. soln.) | |
68399-81-5 | |
White to Yellow | |
Powder | |
Authentic | |
(HOCH2)3CNHCH2CH(OH)CH2SO3H | |
3-(N-Tris-(hydroxymethyl)methylamino)-2-hydroxypropanesulfonic acid | |
RZQXOGQSPBYUKH-UHFFFAOYSA-N | |
3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]-2-hydroxypropane-1-sulfonic acid | |
109334 | |
≥99% (Acid) | |
TAPSO |
(0.1M aq. soln.) | |
220.0°C to 221.0°C | |
4.3 to 5.3 (1% aq. soln. at 25°C) | |
99+% | |
C7H17NO7S | |
MFCD00038352 | |
Solubility in water: ca 129.6g/L (20°C). | |
C(C(CS(=O)(=O)O)O)NC(CO)(CO)CO | |
259.273 | |
259.3 | |
Biochemical |
Safety and Handling
GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- TAPSO
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only