missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Taq CORE Kit 10
Designed for the amplification of specific regions of DNA using PCR. MP Biomedicals Taq CORE Kit 10 provides the enzymes, buffers and dNTPs necessary for amplification, including the (NH4)2SO4 buffer, which has been report
Brand: MP Biomedicals™ 11EPTQS901K
This item is not returnable.
View return policy
Description

Specifications
Freeze at -20°C | |
10 × 250 units | |
Taq DNA Polymerase 5U/μl |
Frozen | |
Standard PCR |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Taq CORE Kit 10 > 10 x 250 units
Spot an opportunity for improvement?Share a Content Correction