missing translation for 'onlineSavingsMsg'
Learn More

MP BIOMEDICALS Taq DNA Polymerase

Recombinant form, for general PCR applications, includes direct loading incubation buffer.

Brand:  MP BIOMEDICALS 11EPTQL025

Product Code. 11450486

  • 165.00 € / 250 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Taq DNA Polymerase Direct Loading Buffer.
  • Taq Conc: 5 U/μL.

Application Notes:

  • Taq DNA Polymerase from Thermus aquaticus is a thermostable DNA polymerase that is used for the DNA polymerase chain reaction (PCR) in order to amplify DNA sequences.

Usage Statement:

  • Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Key Applications:

  • Molecular Biology
  • PCR/Amplification
  • PCR
  • cDNA synthesis
  • RT-PCR
  • DNA labeling
  • Routine PCR
TRUSTED_SUSTAINABILITY
Specifications

Specifications

30 min.
DNA Polymerase
50 μL
5 U/μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.