Learn More
MP Biomedicals™ Taq DNA Polymerase (5U/μL)
Includes Direct Loading Buffer for loading of sample onto agarose gel after amplification
Brand: MP Biomedicals™ 11EPTQL100
Includes: Taq DNA Polymerase, 5X Direct Loading Buffer with Mg, 25mM MgCl2
Description
Taq polymerase catalyzes oligonucleotide primer-driven, DNA template dependent incorporation of dNTPs into complimentary DNA strands. It displays both 5' to 3' polymerase and exonuclease activities. The included 5X Direct Loading buffer contains a densifying agent and a red-purple dye that allows the migration of the sample to be tracked during electrophoresis.
Unit Definition: One unit will incorporate 10nmol of total dNTPs into acid-precipitable DNA in 30 min. at 74°C.

Specifications
Store at -20°C | |
Standard PCR, RT-PCR, Sequencing, Nucleic Acid Labeling | |
Taq DNA Polymerase, 5X Direct Loading Buffer with Mg, 25mM MgCl2 |
1000U | |
5U/μL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.