missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Taq DNA Polymerase (cloned)

For PCR and other procedures requiring DNA polymerase activity at elevated temperatures

Brand:  Cytiva 27-0798-06

615.87 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 10629275

  • 687.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

One unit incorporates 10nmol of total nucleotides into acid-insoluble material in 30 min. at 70°C in a total volume of 50μL.

  • Purity: Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities
  • Concentration: 5000 units/mL
  • Functional testing: Tested in PCR
  • Storgae Buffer: 50mM Tris-HCl (pH 7.5), 5mM DTT, 0.1mM EDTA, 50% glycerol, stabilizers

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100mM Tris HCl, pH 9, 15mM MgCl2 and 500mM KCl. Store at -20°C
Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities.
Standard PCR
7.5
10 x 250U
5000U/mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.