missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Taq DNA Polymerase, native (5 U/μL)

Product Code. p-200086901 Shop All Thermo Scientific Products
Click to view available options
Format:
With BSA
Without BSA
Conditionnement:
500 units
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 10756421

Marque: Thermo Scientific™ EP0072

Please to purchase this item. Need a web account? Register with us today!

Edge
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Thermo Scientific Native Taq DNA Polymerase is a highly thermostable DNA polymerase of the thermophilic bacterium Thermus aquaticus.

Thermo Scientific Taq DNA Polymerase, native, is a highly thermostable DNA polymerase of the thermophilic bacterium Thermus aquaticus. The enzyme catalyzes 5' to 3' synthesis of DNA, has no detectable 3' to 5' exonuclease (proofreading) activity, and possesses low 5' to 3' exonuclease activity. In addition, the polymerase exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Native Taq DNA Polymerase is preferred for amplification of bacterial DNA sequences homologous to those found in E. coli.

Available with or without the stabilizing agent BSA, Taq DNA Polymerase, native, with BSA, is often considered the best choice when amplifying DNA samples of lower purity, e.g., genomic DNA from mouse tail.

Both native Taq DNA polymerases are supplied with two buffers: 10X Taq Buffer with KCl and 10X Taq Buffer with (NH4)2SO4. The latter allows for PCR at a wide range of magnesium concentrations and decreases unspecific priming

Features

  • Thermostable with half-life more than 40 min at 95°C
  • Generates PCR products with 3'-dA overhangs
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescent-labeled nucleotides)

Applications

  • Routine PCR amplification of DNA fragments up to 5 kb
  • DNA labeling
  • High throughput PCR

Notes

  • The error rate of Taq DNA Polymerase in PCR is 2.2 x 10-5 errors per nt per cycle. Accordingly, the accuracy of PCR is 4.5 x 104. Accuracy is an inverse of the error rate and shows an average number of correct nucleotides incorporated before an error occurs.
  • The 10X Taq Buffer without Detergent is recommended for microarray experiments.
TRUSTED_SUSTAINABILITY

Spécification

Concentration 5 U/μL
Content And Storage Taq DNA Polymerase, native (5 U/μL), 100 μL
• 10X Taq Buffer with KCl, 2 x 1.25 mL
• 10X Taq Buffer with (NH4)2SO4, 2 x 1.25 mL
• 25 mM MgCl2, 2 x 1.25 mL
• BSA (20 mg/mL), 0.25 mL

Store at -20°C.
Format With BSA
Polymerase Taq DNA Polymerase
Optimal Reaction Temperature 72°C
Product Type DNA Polymerase
Quantity 500 units
Research Category PCR
Shipping Condition Dry Ice
For Use With (Application) Standard PCR

For Research Use Only. Not for use in diagnostic procedures.

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis