missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Taq DNA Polymerase, recombinant (1 U/μL)

Thermo Scientific Taq DNA Polymerase is a highly thermostable DNA polymerase from the thermophilic bacterium Thermus aquaticus.

Brand:  Thermo Scientific™ EP0404

148.20 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 10180780

  • 228.00 € / 500 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Taq DNA Polymerase is a highly thermostable DNA polymerase from the thermophilic bacterium Thermus aquaticus. The enzyme catalyzes 5' to 3' synthesis of DNA, has no detectable 3' to 5' exonuclease (proofreading) activity, and possesses low 5' to 3' exonuclease activity. In addition, Taq DNA Polymerase exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Recombinant Taq DNA Polymerase is the ideal tool for standard PCR of templates 5 kb or shorter.

Taq DNA Polymerase is supplied with two buffers: 10X Taq Buffer with KCl and 10X Taq Buffer with (NH4)2SO4. The latter allows for PCR at wide range of magnesium concentrations and decreases unspecific priming.

Features

  • High thermostability with half-life greater than 40 min at 95°C
  • Generates PCR products with 3'-dA overhangs
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescent-labeled nucleotides)

Applications

  • Routine PCR amplification of DNA fragments up to 5 kb
  • High throughput PCR
  • DNA labeling

Notes

  • The error rate of Taq DNA Polymerase in PCR is 2.2 x 10-5 errors per nt per cycle. Accordingly, the accuracy of PCR is 4.5 x 104. Accuracy is an inverse of the error rate and shows an average number of correct nucleotides incorporated before an error occurs.
  • 10X Taq Buffer without Detergent is recommended for microarray experiments.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Taq DNA Polymerase (1 U/μL), 500 μL
• 10X Taq Buffer with KCl, 2 x 1.25 mL
• 10X Taq Buffer with (NH4)2SO4, 2 x 1.25 mL
• 25 mM MgCl2, 2 x 1.25 mL

Store at -20°C.
Taq DNA Polymerase
Taq DNA Polymerase (recombinant)
Standard PCR
1X
3'-A
Low
Standard
500 Units
1 U/μL
No
Separate Components
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.