Learn More
MP Biomedicals™ Taq Pol Bulk
For life science applications
Brand:  MP Biomedicals™ 11EPTQA925
 View more versions of this product
Product Code. 11417020
  • 1360.00 € / 250 units
Explore more special offers
Description
Description
  • Ready for CE IVD labeling of clinical applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Taq Pol Bulk
10 x 250U
5U/μL
Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Resaerch Use Only