Learn More
Applied Biosystems™ TaqMan™ Array Human MicroRNA A+B Cards Set v3.0
TaqMan™ Array Human MicroRNA A+B Cards Set v3.0
Brand:  Applied Biosystems™ 4444913
1226.55 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 12007799
  • 2405.00 € / Pack of 8
Explore more special offers
Description
Description
The TaqMan™ Array Human MicroRNA Card Set v3.0 is a two card set containing a total of 384 TaqMan™ MicroRNA Assays per card. The set enables accurate quantitation of 754 human microRNAs in as little as 5 hours. Included on each array is three TaqMan MicroRNA Assay endogenous controls to aid in data normalization and one TaqMan™ MicroRNA Assay not related to human as a negative control. Details on the included assays is available in the Target List file. Purchase of the Megaplex™ RT Primers, Human Pool Set v3.0 is required to run this array set. An additional preamplification step is enabled by Megaplex™ PreAmp Primers, Human Pool Set v3.0 for situations where sensitivity is of the utmost importance and/or sample is limiting.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Array Human MicroRNA A+B Cards Set
Human
Dried Down
FAM
8 pack
Gene Regulation, Epigenetics
384-well Microfluidic Card
4°C
AB 7900 HT 384-Well System
Megaplex™
Standard