Learn More
Applied Biosystems™ TaqMan™ Array, Human Telomere Extension by Telomerase, Fast 96-well
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state.
Brand: Applied Biosystems™ 4418832
119.85 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Description
Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The TaqMan™ Array Human Telomere Extension by Telomerase 96-well Plate contains 28 assays to telomere extension by telomerase associated genes and 4 assays to candidate endogenous control genes. All assays are plated in triplicate.
Specifications
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C. | |
96-well plate | |
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') | |
TaqMan™ | |
Cell Cycle & Proliferation, Development | |
Gene Expression, Oncology, Genetic Disease Research |
Dried Down | |
Fast | |
FAM | |
1 x 96-well plate | |
Human | |
1 x 10 μL Reactions |
For Research Use Only. Not for use in diagnostic procedures.