missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Array Human Transcription of mRNA
Contains 44 assays to transcription of mRNA associated genes and 4 assays to candidate endogenous control genes
Brand: Applied Biosystems™ 4414194
116.28 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
- The Applied Biosystems™ TaqMan™ Array Human Transcription of mRNA 96-well Plate contains:
- 44 assays to transcription of mRNA associated genes
- 4 assays to candidate endogenous control genes
- All assays are plated in duplicate
- Convenient; dried PCR primer and TaqMan™ probe sets supplied in a 96- well plate; just add master mix and your sample
- Same TaqMan™ Assay Quality, New Format
Just Add Master Mix and Sample, and Begin Cycling
- Ideal for projects such as validation of microarray and RNAi data, and pathway studies
- The plates include just the right amount of TaqMan™ Assay in each well: enough for one 20 μl reaction
- You can order as few as three Gene Signature Plates, making these an affordable option for many laboratories and minimizing the waste of unused individually purchased assays
- Save a significant amount of time setting up your experiments
Plate Map Ensures Proper Data Labeling and Easy Re-Ordering
- Each pack of TaqMan™ Array Plates is shipped with a CD containing the plate configuration map, along with the details of each TaqMan™ Assay
- This information is imported into the real-time PCR instrument to enable proper labeling of experimental data and easy access to Assay ID numbers for simple reordering
Simple to Use
- The plates can be stored at room temperature or at 4°C
- When you are ready to run your assays, simply add PCR master mix and your cDNA sample, seal the plate, and begin cycling on a real-time PCR instrument
Compatibility
- TaqMan™ Array Plates have been optimized for use with TaqMan™ Gene Expression Master Mix, but are also compatible with TaqMan™ Universal PCR Master Mix
- For studies using cultured cells, TaqMan™ Array Plates are also compatible (and recommended) with the TaqMan™ Gene Expression Cells-to-Ct™ Kit
- TaqMan™ Array Plates must be run on a real-time PCR instrument with 96-well plate capability
- The plates have been validated for use on Applied Biosystems™ 7000, 7300, 7500 and 7900HT Fast Real-Time PCR Systems using standard 96-well blocks, standard PCR cycling conditions, and 20 μl reaction volumes
Specifications
Array Plate | |
Human | |
Dried Down | |
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') | |
TaqMan™ | |
1 standard plate | |
Gene Regulation |
96-array Plate | |
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C. | |
TaqMan Assay for One 20 uL Reaction | |
FAM | |
Standard | |
Standard |
For Research Use Only. Not for use in diagnostic procedures.