missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Copy Number Reference Assay, human, RNase P
GREENER_CHOICE

Human TaqMan™ Copy Number Reference Assays are run with human TaqMan™ Copy Number Assays in a duplex real-time PCR reaction to detect and measure copy number variations (CNVs) and smaller regions in the human genome.

Brand:  Applied Biosystems™ 4403326

115.90 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10535585

  • 193.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Assays are run with human TaqMan™ Copy Number Assays in a duplex real-time PCR reaction
  • Used to detect and measure copy number variations (CNVs) and smaller regions in the human genome
  • Two options for genes that can be used as endogenous reference genes in humans are offered: RNase P and TERT
  • Designed to unique genomic sequences in the reference genome assembly (build GRCh38) and are required for relative quantitation of copy number targets
  • TaqMan™ Copy Number Assays quantitate the gene of interest and normalize to an endogenous reference gene known to be present in two copies in a diploid genome
  • Please note that TaqMan™ Copy Number Reference Assays are species specific.
  • Assays should be stored at -15°C to -25°C in the dark
  • Minimize freeze-thaw cycles

TaqMan™ Copy Number Reference Assay RNase P

  • Recommended as the standard reference assay for copy number analysis
  • This assay detects the Ribonuclease P RNA component H1 (H1RNA) gene (RPPH1) on chromosome 14, cytoband 14q11.2
  • The assay location is chr.14:20343370 on build GRCh38
  • It has an 87 bp amplicon that maps within the single exon RPPH1 gene

TaqMan™ Copy Number Reference Assay TERT

  • An alternative reference assay recommended in the event that the RNase P assay functions poorly with a sample
  • This could be due to chromosomal aberrations or other issues
  • This assay targets the telomerase reverse transcriptase (TERT) gene located on chromosome 5, cytoband 5p15.33
  • The assay location is chr5:1253257 on build GRCh38
  • It has an 88 bp amplicon that maps within exon 16 of the TERT gene
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1909044 Certificate TypeCertificate of Analysis Date22/07/2020 Catalog Number4403326
Lot Number2002048 Certificate TypeCertificate of Analysis Date22/07/2020 Catalog Number4403326
Lot Number2005050 Certificate TypeCertificate of Analysis Date22/07/2020 Catalog Number4403326
Lot Number1909043 Certificate TypeCertificate of Analysis Date22/07/2020 Catalog Number4403326
Lot Number1912047 Certificate TypeCertificate of Analysis Date22/07/2020 Catalog Number4403326
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ TaqMan™ Copy Number Reference Assay, human, RNase P > 750 reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.