missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Drug Metabolism Genotyping Assay
GREENER_CHOICE

TaqMan™ Drug Metabolism SNP Genotyping Assay

Brand:  Applied Biosystems™ 4362691


Product Code. 10650796

  • 578.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TaqMan™ SNP Genotyping Assays make it easy to perform human and mouse SNP genotyping studies with the precision of TaqMan™ reagent-based chemistry. 4.5 million SNP assays are available, including 3.5 million HapMap SNPs, 70,000 cSNPs and 160,000 validated assays.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 tube containing a 20X mix of pre-formulated assay (2 probes and 2 primers).

Store at -15 to -25°C.

Reagent
TaqMan™
Room Temperature
Assays for 220 drug metabolism and transporter genes are available
20X
FAM, VIC
150 reactions, Inventoried
Human
Frozen
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • TaqMan Drug Metabolism Genotyping Assay

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Formamide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.