missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Fast Reagent Starter Kit
Designed to detect and quantify Eukaryotic 18S rRNA sequence in cDNA samples using the TaqMan 5' nuclease assay, in fast thermal cycling mode.
Brand: Applied Biosystems™
240.30 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
The Eukaryotic 18S rRNA TaqMan™ Assay consists of a 20X mix of two unlabeled PCR primers and a FAM™ dye labeled 18S TaqMan™ MGB probe. Used With: 7900HT Fast Real-Time PCR System with Fast 96-well Block Module, 7900HT Fast Real-Time Service Upgrade, 7900HT System Fast 96-well Block Upgrade Kit.
Order Info
Dry Ice
Specifications
This kit contains TaqMan™ Fast Real-Time PCR Universal Master Mix (2X), no AmpErase™ UNG (1 tube) , Eukaryotic 18S rRNA TaqMan™ Assay (20X mix of two unlabeled PCR primers and a FAM™ dye-labeled 18S TaqMan™ MGB probe) (Assay ID Hs99999901_s1; Part Number 4331182), and human Raji cDNA. Store the PCR mix at 2° to 8°C Store the rest of the kit contents at -15° to -25°C. |
|
qPCR | |
Fast | |
ROX (Pre-mixed) | |
PCR Reagents | |
125 Reactions | |
Target-Specific | |
No | |
DNA (Genomic), cDNA |
Primer-probe | |
AmpliTaq DNA Polymerase | |
7500 Fast System, 7900HT Fast System, AB 7900HT Fast Mode | |
TaqMan™ | |
Dry Ice | |
18S rRNA | |
Gene Expression | |
125 Reactions |
For Research Use Only. Not for use in diagnostic procedures.