missing translation for 'onlineSavingsMsg'
Learn More

TaqMan® TaqMan™ Gene Expression Assay (FAM)
GREENER_CHOICE

Applied Biosystems TaqMan Gene Expression assays are used for quantitative real-time PCR analysis of gene expression and consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM) on the 5' end and a minor groove binder (MGB) and non-fluorescent quencher (NFQ) on the 3' end.

Brand:  TaqMan® 4351368

 View more versions of this product

Product Code. 11997011

  • 1206.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Applied Biosystems TaqMan Gene Expression assays are used for quantitative real-time PCR analysis of gene expression and consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM) on the 5’ end and a minor groove binder (MGB) and non-fluorescent quencher (NFQ) on the 3’ end.

Features of TaqMan Gene Expression assays include:
Easy to use—just add cDNA and master mix and run the qPCR—no melt curves required
Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
Accurate—identify small fold-changes with high accuracy of quantitation
Extensive content—Over 1.8 million predesigned assays available for over 25 different species
Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for gene variants with high sequence homology
Fast delivery—your assays shipped in as little as 1–2 days (see below)

Approximate ship time
Inventoried: 1–2 days in North America, 3–5 days in Europe*
Made to order: 4–6 days in North America, 6–10 days in Europe

TaqMan Gene Expression assays are the gold standard in real-time PCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents.

Pre-formulated assay
• 2 unlabeled PCR primers (900 nM each final 1X concentration)
• 1 FAM dye-labeled TaqMan MGB probe (250 nM final 1X concentration)

All of our predesigned TaqMan Gene Expression assays are covered by the TaqMan Assays qPCR Guarantee.**

Recommended master mix: TaqMan Fast Advanced Master Mix (Cat. No. 4444556)

*Our most popular assays are stocked in North America and Europe for overnight delivery where possible. If an assay is out of stock, it may take an additional 3–5 business days for delivery.

**Subject to terms and conditions. For complete details, go to www.thermofisher.com/taqmanguarantee.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 tube containing 20X (XS, S, and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15°C to -25°C.

Tube
qPCR
7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
TaqMan™
L (2900 reactions/967 μL), made to order
60X
L (2900 x 967 μL Reactions), made to order
Primer-probe
High
Fast
Sustainable packaging
Gene Expression Assay
Room Temperature
60 X
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.