missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Protein Assays Core Reagents Kit with MasterMix

Contains both the Core Reagents Base Kit and the TaqMan Protein Assays Fast Master Mix Kit.

Brand:  Applied Biosystems™ 4448591

Product Code. 12037799

  • 2020.00 € / 500 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Core Reagents Base Kit contains the reagents required to perform the ligation and PCR amplification steps of TaqMan™ protein expression experiments. The Universal PCR Assay contains optimized concentrations of TaqMan™ probe and oligonucleotide primers (forward and reverse) that are designed to amplify a region of the protease-treated ligation product for detection on an Applied Biosystems real-time PCR system. The enzyme concentration, ionic strength, and the pH of the TaqMan™ Protein Expression Fast Master Mix are optimized for the amplification of ligated oligonucleotide target sequences prepared using TaqMan™ Protein Expression Assay reagents. The DNA polymerase included in the master mix is designed for maximum PCR efficiency and specificity when used with the ligation chemistry.

  • PCR Method: qPCR Protein Quantification
  • Primer-Probe Compatability: TaqMan™
TRUSTED_SUSTAINABILITY
Specifications

Specifications

The TaqMan™ Protein Assays Core Reagents Kit with MasterMix contains both the Core Reagents Base Kit and the TaqMan Protein Assays Fast Master Mix Kit.
qPCR
Fast
7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOnePlus™, Fast Mode
ROX (Pre-mixed)
Protein Assays Core Reagent Kit
500 Reactions
No
Tissue, Cells
Primer-probe
AmpliTaq Fast DNA Polymerase
Proximity Ligation Assay
FAM
TaqMan™
500 reactions
Not Target-Specific
500 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ TaqMan™ Protein Assays Core Reagents Kit with MasterMix > 500 Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.