missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Protein Expression Core Reagents Kit
TaqMan™ Protein Expression Core Reagents Kit
Brand: Applied Biosystems™ 4405501
328.44 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Applied Biosystems™ TaqMan™ Protein Expression Assays enable fast, easy identification, and relative protein quantification of pluripotency protein markers from limited quantities of cultured hESCs. The TaqMan™ Protein Expression Assays Core Reagents Base Kit contains the reagents required to perform the ligation and PCR amplification steps of TaqMan™ protein expression assays. The Universal PCR Assay contains optimized concentrations of TaqMan™ probe and oligonucleotide primers (forward and reverse) that are designed to amplify a region of the protease-treated ligation product for detection on an Applied Biosystems™ real-time PCR system. The enzyme concentration, ionic strength, and the pH of the TaqMan™ Protein Expression Fast Master Mix are optimized for the amplification of ligated oligonucleotide target sequences prepared using TaqMan™ Protein Expression Assay reagents. The DNA polymerase included in the master mix is designed for maximum PCR efficiency and specificity when used with the ligation chemistry. Key features of the TaqMan™ Protein Expression Assay:Specifications
Includes the TaqMan™ protein expression core reagents base kit (with DNA ligase, ligase dilution buffer, ligation reaction buffer, PBS pH 7.4, protease, and universal PCR assay; store at –15 to –25°C), and the TaqMan™ protein expression fast master mix (store at 2 to 8°C) | |
7500 Fast system, 7900HT fast system, StepOne™, fast mode, StepOnePlus™, fast mode | |
TaqMan™ Protein Expression Core Reagents Kit | |
100 rxn(s) | |
Protein Expression Analysis, Stem Cell Analysis | |
100 rxn(s) |
Primer-probe | |
FAM | |
UP (Ultra Pure) | |
Not target-specific | |
TaqMan™ Protein Expression Core Reagents kit | |
Cells (general) |
For Research Use Only. Not for use in diagnostic procedures.