missing translation for 'onlineSavingsMsg'
Learn More

TaqMan™, RNA-to-CT™ TaqMan™ RNA-to-CT1-Step Kit

Delivers consistent RNA target quantification for a wide variety of assays and is validated with Applied Biosystems™ comprehensive library of TaqMan™ Gene Expression assays.

Brand:  TaqMan™, RNA-to-CT™ 4392653

 View more versions of this product

Product Code. 10158014

  • 315.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The TaqMan™ RNA-to-CT™ One-Step Kit is recommended for a variety of applications including general gene expression studies, biomarker discovery, and microarray validation.

  • Accurate across a wide dynamic range for dependable performance
  • High sensitivity, enabling low copy number detection and precise quantification
  • Validated with TaqMan™ Gene Expression Assays for easy experimental setup
  • Consistent performance with a wide variety of targets, including AT-rich, GC-rich and long sequences
  • Fewer pipetting steps means less hands-on-time and less chance for errors

Efficiency across a Broad Range of Input Concentrations
  • Maximum flexibility in RNA template input quantity requires optimal real-time PCR efficiency across a broad range of template quantities
  • The result is comparable to that of the TaqMan™ RNA-to-CT™ 2-Step Kit, and is a significant improvement in dynamic range and sensitivity over the TaqMan™ One-Step RT-PCR Master Mix Reagents.

High Sensitivity at Low Target Concentration
  • The sensitivity of TaqMan™ RNA-to-CT™ One-Step Kit was validated using a synthetic RNA template for which copy number is precisely known
  • Significant sampling error occurs when measuring low quantities of target, so statistical analysis is required for proper evaluation using multiple replicates
  • Statistical analysis indicates high confidence of sample quantification based on a T-test, consistent with as few as 10 copies of target.

Order Info

Shipping Conditions: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sufficient for 40 reactions at 50 μL reaction volume. Includes:
• 1 x 1 mL tube of 2X master mix containing AmpliTaq Gold™ DNA Polymerase UP, dNTPs (including dUTP), Passive Reference 1, and optimized buffer components. Store at -15 to -25°C.
• 1 x 50 μL tube of 40X RT enzyme mix containing ArrayScript™ UP Reverse Transcriptase, RNase Inhibitor. Store at -15° to -30°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
1-step RT-qPCR
Standard
ROX (Pre-mixed)
One-Step RT-PCR Kit
40 reactions
Wet Ice
Gene Expression
RNA
Primer-probe
AmpliTaq Gold DNA Polymerase
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
RNA-to-CT™, TaqMan™
UP (Ultra Pure)
ArrayScript™ UP
40 Reactions
40 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.