missing translation for 'onlineSavingsMsg'
Learn More

TaqMan™, RNA-to-CT™ TaqMan™ RNA-to-CT1-Step Kit

Product Code. 10734807 Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
200 Reactions
2000 Reactions
40 Reactions
Unit Size:
1 set
200 reactions
Each
This item is not returnable. View return policy

Product Code. 10734807

Brand: TaqMan™, RNA-to-CT™ 4392938

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Delivers consistent RNA target quantification for a wide variety of assays and is validated with Applied Biosystems™ comprehensive library of TaqMan™ Gene Expression assays.

The TaqMan™ RNA-to-CT™ One-Step Kit is recommended for a variety of applications including general gene expression studies, biomarker discovery, and microarray validation.

  • Accurate across a wide dynamic range for dependable performance
  • High sensitivity, enabling low copy number detection and precise quantification
  • Validated with TaqMan™ Gene Expression Assays for easy experimental setup
  • Consistent performance with a wide variety of targets, including AT-rich, GC-rich and long sequences
  • Fewer pipetting steps means less hands-on-time and less chance for errors

Efficiency across a Broad Range of Input Concentrations
  • Maximum flexibility in RNA template input quantity requires optimal real-time PCR efficiency across a broad range of template quantities
  • The result is comparable to that of the TaqMan™ RNA-to-CT™ 2-Step Kit, and is a significant improvement in dynamic range and sensitivity over the TaqMan™ One-Step RT-PCR Master Mix Reagents.

High Sensitivity at Low Target Concentration
  • The sensitivity of TaqMan™ RNA-to-CT™ One-Step Kit was validated using a synthetic RNA template for which copy number is precisely known
  • Significant sampling error occurs when measuring low quantities of target, so statistical analysis is required for proper evaluation using multiple replicates
  • Statistical analysis indicates high confidence of sample quantification based on a T-test, consistent with as few as 10 copies of target.

Order Info

Shipping Conditions: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains sufficient reagents for 200 reactions at 50 μl reaction volumes. Includes:
• 1 × 5 ml tube of 2X master mix containing AmpliTaq Gold™ DNA Polymerase UP, dNTPs (including dUTP), Passive Reference 1, and optimized buffer components. Store at -15 to -25°C.
• 1 × 250 μl tube of 40X RT enzyme mix containing ArrayScript™ UP Reverse Transcriptase, RNase Inhibitor. Store at -15°C to -30°C.
Detection Method Primer-probe
PCR Method 1-step RT-qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
Passive Reference Dye ROX (Pre-mixed)
Product Line RNA-to-CT, TaqMan
Product Type One-Step RT-PCR Kit
Purity or Quality Grade UP (Ultra Pure)
Quantity 200 reactions
Reverse Transcriptase ArrayScript™ UP
Shipping Condition Wet Ice
Sufficient For 200 Reactions
For Use With (Application) Gene Expression
No. of Reactions 200 Reactions
Sample Type RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.