missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ RNase P Instrument Verification Plate, 96-well

The TaqMan RNase P Instrument Verification Plate is used to verify the performance of the Applied Biosystems 7900HT 96-Well Real-Time PCR System and the ABI PRISM 7000 and 7700 Sequence Detection Systems.

Brand:  Applied Biosystems™ 4310982

Additional Details : Weight : 0.13000kg

Product Code. 10115874

  • 1308.00 € / Each
Estimated Shipment: 12-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The TaqMan RNase P Instrument Verification Plate is used to verify the performance of the Applied Biosystems 7900HT 96-Well Real-Time PCR System and the ABI PRISM 7000 and 7700 Sequence Detection Systems. The TaqMan RNase P Instrument Verification Plate is a 96-well PCR plate preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each well contains preloaded reaction mix (1X TaqMan Universal PCR Master Mix, RNase P primers, and FAM™ dye–labeled probe) and template.

The TaqMan RNase P Instrument Verification Plate satisfies the same requirements as the Applied Biosystems 7900HT 96-Well Real-Time PCR System, and the ABI PRISM 7000 and 7700 Sequence Detection Systems installations. To pass installation, the instruments must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, Applied Biosystems recommends that the performance of your instrument be verified every six months.

Specifications

Specifications

96-well
FAM
Fluorescence
Instrument Verification Plate
Dry Ice
RNase P Calibration, Instrument Verification Plate
Contains a 96-well Reaction Plate pre-loaded and sealed with the 5' nuclease assay using RNaseP TaqMan™ reagents, including primers and probes.

Store at -15°C to -25°C. Protect from light.
7000 System, 7700 System, 7900HT System
TaqMan™
Real Time PCR (qPCR)
1 x 96-well Plate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.