missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TASP 0390325

Potent and selective V1B receptor antagonist; orally bioavailable

297.00 € - 1170.00 €

Specifications

Product Type Antagonist
Target Proteins Vasopressin Receptor
CAS 1642187-96-9
Molecular Formula C25H29ClFN5O4.HCl
Formula Weight 554.44g/mol
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18770258
View Documents
Tocris Bioscience™
5729/10
10 mg
297.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
18720118
View Documents
Tocris Bioscience™
5729/50
50 mg
1170.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potent and selective V1B receptor antagonist (IC50 = 2.22 nM for rat pituitary receptor). Exhibits no significant affinity for 85 other receptors, ion channels and transporters at 10 μM, including V1A and V2. Increases ACTH levels in rats. Orally bioavailable. Antidepressant and anxiolytic.
Specifications

Specifications

Antagonist
1642187-96-9
554.44g/mol
Vasopressin Receptor Antagonists
Vasopressin Receptor
C25H29ClFN5O4.HCl
Store at -20°C
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only