Learn More
Taurine, 99%, Thermo Scientific Chemicals
Description
Taurine plays an important role in cardiovascular function, skeletal muscle function, the retina and the central nervous system. It acts as an antioxidant as well as protecting against the toxicity of various substances such as lead and cadmium. It is also used in contact lens solution and an ingredient in numerous energy drinks.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Taurine plays an important role in cardiovascular function, skeletal muscle function, the retina and the central nervous system. It acts as an antioxidant as well as protecting against the toxicity of various substances such as lead and cadmium. It is also used in contact lens solution and an ingredient in numerous energy drinks.
Solubility
Soluble in water and dimethyl sulfoxide. Insoluble in absolute ethanol and diethyl ether.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | >300°C |
Assay Percent Range | 99% |
Quantity | 100 g |
Beilstein | 1751215 |
Solubility Information | Soluble in water and dimethyl sulfoxide. Insoluble in absolute ethanol and diethyl ether. |
Formula Weight | 125.15 |
Chemical Name or Material | Taurine |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.