Learn More
Thermo Scientific Chemicals TBE (10X), TRIS + borate + EDTA
49.95 € - 139.70 €
Specifications
Chemical Name or Material | Buffer |
---|---|
Description | Colorless |
Concentration | 10X |
Physical Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15424829
|
Thermo Scientific Chemicals
J62449.K2 |
1 L |
49.95 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15434829
|
Thermo Scientific Chemicals
J62449.K3 |
2 L |
77.18 €
2 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15444829
|
Thermo Scientific Chemicals
J62449.K7 |
4 L |
139.70 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TBE (Tris/Borate/EDTA) Buffer is commonly used in nucleic acid electrophoresis. TBE Buffer is used for polyacrylamide and agarose gel electrophoresis. It is ideally suited for DNA and RNA separation on longer run, higher voltage or amperage gels. TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel.5 TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids. Tris-Borate-EDTA buffer has been used for pulsed-field gel electrophoresis (PFGE).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer | |
Colorless | |
10X | |
Liquid |
Safety and Handling
Recommended Storage : Ambient temperatures
- TBE (10X), TRIS + borate + EDTA
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Boric acid (H3BO3)
RUO – Research Use Only