missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™
Brand: Fisher BioReagents BP2471-1
42.83 EUR valid until 2024-12-31
Use promo code "21951" to get your promotional price.
This item is not returnable.
View return policy
Description
TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology
- Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X TBS buffer solution - simply dilute as needed
- 10X solution contains 1.37M NaCl, 0.027M KCl and 0.25M tris
- To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TBS solution with 9 volumes of ultrapure water, such as BP2819
- Filtered and autoclaved - ready to use stock TBS buffer
- Tested for the absence of DNase, RNase, and protease
- TBS buffer is often used in immunodetection techniques such as Western blotting protocols
- Intended for laboratory research use only
Specifications
Tris Buffered Saline | |
7447-40-7, 7647-01-0, 7647-14-5, 77-86-1, 7732-18-5 | |
Colorless | |
Liquid | |
DNase free | |
Nalgene Poly Bottle | |
TBS | |
10X solution contains 1.37M Sodium Chloride, 0.027M Potassium Chloride, and 0.25M Tris/Tris-HCl. |
10X Solution | |
100°C | |
7.5 | |
1 L | |
DNase-, RNase- and Protease-Free | |
Protease free | |
Molecular Biology |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction