missing translation for 'onlineSavingsMsg'
Learn More

TBS with Tween™ (TBST), 20X Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Used in Western blotting protocols

183.00 € - 348.00 €

Specifications

Product Type TBS with Tween (TBST)
Form Solution
Concentration 20X
Content And Storage Ambient temperatures
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15805428
View Documents
Thermo Scientific
J77500.K2
1 L
183.00 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15815428
View Documents
Thermo Scientific
J77500.K8
5 L
348.00 €
5 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used in Western blotting protocols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

TBS with Tween (TBST)
Solution
20X
Ambient temperatures
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.