missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TC-100 Insect Medium
GREENER_CHOICE

TC-100 Insect Medium

Brand:  Gibco™ 13055025

Product Code. 11510456

  • 75.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

TC 100 Insect Medium was originally designed for the growth of insect cells, including Spodeptera frugiperda (Sf9), and the expression of recombinant Autographa Californica Nuclear Polyhedrosis Virus (rAcNPV) using the Baculovirus Expression Vector System (BEVS). We offer a variety of Gibco™ Media options for a range of insect cell culture applications. TC 100 Insect Medium is rich in amino acids and uses tryptose as an additional sugar source.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

cGMP Manufacturing and Quality System
Gibco™ TC 100 Insect Medium is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Gibco™ TC 100 Insect Medium is commonly supplemented with 10% Fetal Bovine Serum (FBS) before use. TC 100 Insect Medium requires ambient CO2 in order to maintain physiological pH.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Insect Cell
Liquid
Glutamine, Tryptose Phosphate Broth, Sodium Bicarbonate
500 mL
Sf21, Sf9
Insect Cell Culture
Insect Cell Medium
Gibco™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.