missing translation for 'onlineSavingsMsg'
Learn More
TC ASK 10, Tocris Bioscience™ Product Code.: 30001633

TC ASK 10, Tocris Bioscience™

Product Code. 30001633
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 30001633

Brand: Tocris Bioscience 4825/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective ASK1 inhibitor; orally bioavailable

Potent ASK1 inhibitor (IC50 = 14 nM); displays selectivity for ASK1 over other kinases including ASK2 (IC50 = 0.51 μM), MEKK1, TAK1, IKKβ, ERK1, JNK1, p38α, GSK-3β, PKCθ and B-raf (IC50 values are > 10 μM). Blocks downstream JNK1/p38 phosphorylation in cells. Orally bioavailable.

Chemical Identifiers

CAS 1005775-56-3
Molecular Formula C21H23Cl2N5O
Molecular Weight (g/mol) 432.349
InChI Key IKKLFEDUYFZNBO-UHFFFAOYSA-N
Synonym ask1 inhibitor 10, tc ask 10, 4-tert-butyl-n-6-1h-imidazol-1-yl imidazo 1,2-a pyridin-2-yl benzamide dihydrochloride, 4-1,1-dimethylethyl-n-6-1h-imidazol-1-yl imidazo 1,2-a pyridin-2-yl benzamide dihydrochlorideShow MoreShow Less
PubChem CID 68661090
IUPAC Name 4-tert-butyl-N-(6-imidazol-1-ylimidazo[1,2-a]pyridin-2-yl)benzamide;dihydrochloride
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)NC2=CN3C=C(C=CC3=N2)N4C=CN=C4.Cl.Cl

Specifications

Quantity 50 mg
Formula Weight 432.35
Percent Purity >98%
Chemical Name or Material TC ASK 10
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TC ASK 10, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.