Learn More
HUBER TC100E-F Model Stainless Steel Immersion Cooler
Used for rapid cooling of fluids.
Brand:  HUBER 3005.0046.99
Product Code. 10226433
  • 14580.00 € / Each
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Rapid cooling of fluids;
  • Ideal for counter-cooling or as a replacement of dry ice
  • LED temperature display
  • Easy operation with only three keys
  • Compact, space-saving construction
  • Robust housing with carrying grips
  • High quality stainless steel immersion probe
  • Temperature controller, stability ±0.5°C
Specifications
Specifications
-100°C to +40°C
TC100E-F
220/240 V