missing translation for 'onlineSavingsMsg'
Learn More

TCN 201, Tocris Bioscience™

Product Code. 18014562
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

GluN1/GluN2A-selective NMDA antagonist

NMDA receptor antagonist selective for GluN1/GluN2A (formally NR1/NR2A) over GluN1/GluN2B (formally NR1/NR2B) containing receptors (pIC50 values are 6.8 and <4.3, respectively, in human recombinant GluN1/GluN2A (formally NR1/NR2A) and GluN1/GluN2B (formally NR1/NR2B) FLIPR/Ca2+ assays).

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 852918-02-6
Molecular Formula C21H17ClFN3O4S
Molecular Weight (g/mol) 461.892
InChI Key FYIBXBFDXNPBSF-UHFFFAOYSA-N
Synonym qcr-22, n-4-2-benzoylhydrazinecarbonyl benzyl-3-chloro-4-fluorobenzenesulfonamide, 3-chloro-4-fluoro-n-4-2-phenylcarbonyl hydrazino carbonyl benzyl benzenesulfonamide, n-4-benzamidocarbamoyl phenyl methyl-3-chloro-4-fluorobenzenesulfonamide, d09hxx, c21h17clfn3o4so6, tcn hplc, 3-chloro-4-fluoro-n-4-phenylformohydrazido carbonyl phenyl methyl benzene-1-sulfonamide, 4-3-chloro-4-fluorophenyl sulfonyl amino methyl benzoic acid 2-benzoylhydrazide, n-4-2-benzoylhydrazine-1-carbonyl benzyl-3-chloro-4-fluorobenzenesulfonamide
PubChem CID 4787937
IUPAC Name N-[[4-(benzamidocarbamoyl)phenyl]methyl]-3-chloro-4-fluorobenzenesulfonamide
SMILES C1=CC=C(C=C1)C(=O)NNC(=O)C2=CC=C(C=C2)CNS(=O)(=O)C3=CC(=C(C=C3)F)Cl

Specifications

Quantity 10 mg
Formula Weight 461.89
Percent Purity >99%
Chemical Name or Material TCN 201
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'