missing translation for 'onlineSavingsMsg'
Learn More
TCS 2002, Tocris Bioscience™ Product Code.: 18054642

TCS 2002, Tocris Bioscience™

Product Code. 18054642
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18054642

Brand: Tocris Bioscience 3869/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent GSK-3β inhibitor

Potent inhibitor of GSK-3β (IC50 = 35 nM). Oral adminstration inhibits cold water stress-induced tau hyperphosphorylation in the mouse brain.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1005201-24-0
Molecular Formula C18H14N2O3S
Molecular Weight (g/mol) 338.381
InChI Key RCKYSTKYIVULEK-UHFFFAOYSA-N
Synonym 2-3-4-methanesulfinylphenyl-1-benzofuran-5-yl-5-methyl-1,3,4-oxadiazole, mmbo, 1,3,4-oxadiazole, 2-methyl-5-3-4-methylsulfinyl phenyl-5-benzofuranyl, 1,3,4-oxadiazole,2-methyl-5-3-4-methylsulfinyl phenyl-5-benzo b furanyl, 2-3-4-methanesulfinyl phenyl-1-benzofuran-5-yl-5-methyl-1,3,4-oxadiazole, 2-methyl-5-3-4-methylsulfinyl phenyl-1-benzofuran-5-yl-1,3,4-oxadiazole, 2-methyl-5-3-4-methylsulfinyl phenyl-5-benzofuranyl-1,3,4-oxadiazoleShow MoreShow Less
PubChem CID 24855958
IUPAC Name 2-methyl-5-[3-(4-methylsulfinylphenyl)-1-benzofuran-5-yl]-1,3,4-oxadiazole
SMILES CC1=NN=C(O1)C2=CC3=C(C=C2)OC=C3C4=CC=C(C=C4)S(=O)C

Specifications

Quantity 10 mg
Formula Weight 338.38
Percent Purity >99%
Chemical Name or Material TCS 2002
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TCS 2002, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.