missing translation for 'onlineSavingsMsg'
Learn More

TCS 2314, Tocris Bioscience™

Product Code. 18035322
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

α4β1 (VLA-4) antagonist

Integrin very late antigen-4 (VLA-4; α4β1) antagonist (IC50 = 4.4 nM). Blocks the activation of inflammatory cells.

Chemical Identifiers

CAS 317353-73-4
Molecular Formula C28H34N4O6
Molecular Weight (g/mol) 522.602
InChI Key ITXAAOWFOURIHK-DEOSSOPVSA-N
Synonym d08xws, piperidinylacetic acid derivative, 1, 1-3s-4-2-4-2-methylphenyl amino carbonyl amino phenyl acety l-3-morpholinyl carbonyl-4-piperidinediacetic acid, 1-3s-4-4-3-2-methylphenyl ureido phenylacetyl morpholine-3beta-yl carbonyl piperidine-4-acetic acid, 2-1-3s-4-2-4-2-methylphenyl carbamoyl amino phenyl acetyl morpholin-3-yl carbonyl piperidin-4-yl acetic acid, 2-1-3s-4-2-4-2-methylphenyl carbamoylamino phenyl acetyl morpholine-3-carbonyl piperidin-4-yl acetic acid
PubChem CID 11226207
IUPAC Name 2-[1-[(3S)-4-[2-[4-[(2-methylphenyl)carbamoylamino]phenyl]acetyl]morpholine-3-carbonyl]piperidin-4-yl]acetic acid
SMILES CC1=CC=CC=C1NC(=O)NC2=CC=C(C=C2)CC(=O)N3CCOCC3C(=O)N4CCC(CC4)CC(=O)O

Specifications

Quantity 10 mg
Formula Weight 522.59
Percent Purity 98%
Chemical Name or Material TCS 2314
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.