missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TDZD 8

Selective non-ATP competitive inhibitor of GSK 3β

289.00 € - 1135.00 €

Specifications

Color White
Product Type Inhibitor
CAS 327036-89-5
Molecular Formula C10H10N2O2S
Formula Weight 222.26g/mol
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18719508
View Documents
Tocris Bioscience™
6092/10
10 mg
289.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
18781799
View Documents
Tocris Bioscience™
6092/50
50 mg
1135.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Selective non-ATP competitive inhibitor of GSK 3β (IC50 = 2 μM); thiadiazolidinone deriviative. Does not inhibit Cdk-1/cyclin B, CK-II, PKA or PKC at >100 μM. Reduces severity of L-dopa-induced dyskinesia in a Parkinson's disease in vivo model.
Specifications

Specifications

White
327036-89-5
222.26g/mol
Soluble to 100mM in DMSO, 100mM in ethanol
Inhibits GSK 3β
Inhibitor
C10H10N2O2S
NP 01139
Store at -20°C
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only